2-Octadecyl-1,4-dimethoxybenzene

Supplier: TCI America

Synonyms: Octadecylhydroxyquinone Dimethyl Ether

O0217-5G
TCO0217-005GEA 275 USD
TCO0217-005G
2-Octadecyl-1,4-dimethoxybenzene
2-Octadecyl-1,4-dimethoxybenzene

CAS Number: 88702-69-6
MDL Number: MFCD00142955
Molecular Formula: C26H46O2
Molecular Weight: 390.65
Form: Crystal
Color: White
Melting point (°C): 54


Other Product Names / Synonyms: Octadecylhydroxyquinone Dimethyl Ether

Formula: C₂₆H₄₆O₂
MW: 390.65 g/mol
Melting Pt: 54 °C
MDL Number: MFCD00142955
CAS Number: 88702-69-6
https://us.vwr.com/stibo/web/std.lang.all/04/96/28990496.jpg

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR