3-Octylthiophene ≥98.0%

Supplier: TCI America

O0213-25G O0213-5G O0213-1G
TCO0213-001GEA 27.41 USD
TCO0213-001G TCO0213-25G TCO0213-005G
3-Octylthiophene ≥98.0%
3-Octylthiophene

CAS Number: 65016-62-8
MDL Number: MFCD00085281
Molecular Formula: C12H20S
Molecular Weight: 196.35
Purity/Analysis Method: 98.0% (GC)
Form: Clear Liquid
Boiling point (°C): 106
Specific Gravity (20/20): 0.92

Formula: C₁₂H₂₀S
MW: 196.36 g/mol
Boiling Pt: 106 °C
Density: 0.92 (20 °C)
MDL Number: MFCD00085281
CAS Number: 65016-62-8
https://us.vwr.com/stibo/web/std.lang.all/04/92/28990492.jpg

Order Now

Specification Test Results

Purity/Analysis Method 98.0% (GC)

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR