1,1'-Oxalyldiimidazole ≥95.0% (by titrimetric analysis)
Supplier: TCI America
Synonyms:
1,2-Di(1H-imidazol-1-yl)ethane-1,2-dione
O0200-5G
O0200-1G
TCO0200-001GEA
57.84
USD
TCO0200-001G
TCO0200-005G
1,1'-Oxalyldiimidazole ≥95.0% (by titrimetric analysis)
1,1'-Oxalyldiimidazole
CAS Number: 18637-83-7
MDL Number: MFCD00043067
Molecular Formula: C8H6N4O2
Molecular Weight: 190.16
Purity/Analysis Method:
Form: Crystal
Formula:
C₈H₆N₄O₂ MW: 190.16 g/mol |
MDL Number:
MFCD00043067 CAS Number: 18637-83-7 |
Specification Test Results
Purity/Analysis Method |
|
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...