1,2,3,4,5,6,7,8-Octahydroacridine ≥97.0% (by GC, titration analysis)

Supplier: TCI America

O0197-5G
TCO0197-005GEA 289 USD
TCO0197-005G
1,2,3,4,5,6,7,8-Octahydroacridine ≥97.0% (by GC, titration analysis)
1,2,3,4,5,6,7,8-Octahydroacridine

CAS Number: 1658-08-8
MDL Number: MFCD00005032
Molecular Formula: C13H17N
Molecular Weight: 187.29
Purity/Analysis Method: 97.0% (GC,T)
Form: Crystal
Melting point (°C): 70

Formula: C₁₃H₁₇N
Melting Pt: 70 °C
MDL Number: MFCD00005032
CAS Number: 1658-08-8
https://us.vwr.com/stibo/web/std.lang.all/04/79/28990479.jpg

Order Now

Specification Test Results

Purity/Analysis Method 97.0% (GC,T)

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR