1,4-Octadiene (cis- and trans- mixture)
O0170-1ML
TCO0170-001MLEA
468.65
USD
TCO0170-001ML
1,4-Octadiene (cis- and trans- mixture)
1,4-Octadiene (cis and trans mixture)
CAS Number: 5675-25-2
MDL Number: MFCD00027235
Molecular Formula: C8H14
Molecular Weight: 110.20
Form: Clear Liquid
Color: Colorless
Specific Gravity (20/20): 0.74
Formula:
C₈H₁₄ Density: 0,74 (20 °C) |
MDL Number:
MFCD00027235 CAS Number: 5675-25-2 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...