Calcium mesoxalate trihydrate ≥98.0% (by titrimetric analysis)

Supplier: TCI America

Synonyms: Mesoxalic Acid Calcium SaltCalcium Ketomalonate

O0076-5G
TCO0076-005GEA 61.9 USD
TCO0076-005G
Calcium mesoxalate trihydrate ≥98.0% (by titrimetric analysis)
Calcium mesoxalate trihydrate

CAS Number: 21085-60-9
MDL Number: MFCD00150291
Molecular Formula: C3H4O6
Molecular Weight: 174.12
Purity/Analysis Method: 98.0% (T)
Form: Crystal


Other Product Names / Synonyms: Mesoxalic Acid Calcium Salt, Calcium Ketomalonate

Formula: C₃H₄O₆
MW: 174.12 g/mol
MDL Number: MFCD00150291
CAS Number: 21085-60-9

Order Now

Specification Test Results

Purity/Analysis Method 98.0% (T)

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR