1,2,3,4,5,6,7,8-Octahydrophenanthrene ≥98.0%

Supplier: TCI America

O0021-5G O0021-1G
TCO0021-001GEA 237.53 USD
TCO0021-001G TCO0021-5G
1,2,3,4,5,6,7,8-Octahydrophenanthrene ≥98.0%
1,2,3,4,5,6,7,8-Octahydrophenanthrene

CAS Number: 5325-97-3
MDL Number: MFCD00058943
Molecular Formula: C14H18
Molecular Weight: 186.30
Purity/Analysis Method: 98.0% (GC)
Form: Clear Liquid
Boiling point (°C): 295
Specific Gravity (20/20): 1.03

Formula: C₁₄H₁₈
Boiling Pt: 295 °C
Density: 1.03 (20 °C)
MDL Number: MFCD00058943
CAS Number: 5325-97-3
https://us.vwr.com/stibo/web/std.lang.all/03/33/28990333.jpg

Order Now

Specification Test Results

Purity/Analysis Method 98.0% (GC)

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR