9-Nitrocamptothecin ≥98.0% (by HPLC)

Supplier: TCI America

Synonyms: Rubitecan

N0822-100MG
TCN0822-100MGEA 303 USD
TCN0822-100MG
9-Nitrocamptothecin ≥98.0% (by HPLC)
9-Nitrocamptothecin

CAS Number: 91421-42-0
MDL Number: MFCD06656294
Molecular Formula: C20H15N3O6
Molecular Weight: 393.36
Purity/Analysis Method: 98.0% (HPLC)
Form: Crystal
Melting point (°C): 268
Specific rotation [a]20/D: 39 deg (C=0.4, CHCl3:MeOH=8:2)


Product Categories: Bioscience, Antitumor Ingredients, Cancer Research Reagents, Quinoline Alkaloids
Other Product Names / Synonyms: Rubitecan

Formula: C₂₀H₁₅N₃O₆
MW: 393.36 g/mol
Melting Pt: 268 °C
MDL Number: MFCD06656294
CAS Number: 91421-42-0
https://us.vwr.com/stibo/web/std.lang.all/00/18/28990018.jpg

Order Now

Specification Test Results

Purity/Analysis Method 98.0% (HPLC)

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR