(R)-(+)-1-(2-Naphthyl)ethanol ≥98.0%

Synonyms: (R)-(+)-alpha-Methyl-2-naphthalenemethanol(R)-1-(Naphthalen-2-yl)ethanol(R)-(+)-2-(1-Hydroxyethyl)naphthalene

N0784-1G
TCN0784-1GEA 940.39 USD
TCN0784-1G
(R)-(+)-1-(2-Naphthyl)ethanol ≥98.0%
(R)-(+)-1-(2-Naphthyl)ethanol

CAS Number: 52193-85-8
MDL Number: MFCD00075344
Molecular Formula: C12H12O
Molecular Weight: 172.23
Purity/Analysis Method: 98.0% (GC)
Form: Crystal
Melting point (°C): 72
Specific rotation [a]20/D: 40 deg (C=1, MeOH)


Product Categories: Synthetic Organic Chemistry, Chiral Building Blocks
Other Product Names / Synonyms: (R)-(+)-alpha-Methyl-2-naphthalenemethanol, (R)-(+)-2-(1-Hydroxyethyl)naphthalene

Formula: C₁₂H₁₂O
MW: 172.23 g/mol
Melting Pt: 72 °C
MDL Number: MFCD00075344
CAS Number: 52193-85-8
https://us.vwr.com/stibo/web/std.lang.all/99/86/28989986.jpg

Order Now

Specification Test Results

Purity/Analysis Method 98.0% (GC)

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR