Allyl[3-(2-nitrobenzenesulfonamido)propyl]carbamate ≥98.0% (by HPLC)

Supplier: TCI America

Synonyms: [3-(2-Nitrobenzenesulfonamido)propyl]carbamic acid Allyl Ester[3-(2-Nitrobenzenesulphonamido)propyl]carbamic acid Allyl Ester

N0717-1G
TCN0717-1GEA 279 USD
TCN0717-1G
Allyl[3-(2-nitrobenzenesulfonamido)propyl]carbamate ≥98.0% (by HPLC)
Allyl[3-(2-nitrobenzenesulfonamido)propyl]carbamate

CAS Number: 312283-45-7
Molecular Formula: C13H17N3O6S
Molecular Weight: 343.35
Purity/Analysis Method: 98.0% (HPLC)
Form: Crystal


Product Categories: Synthetic Organic Chemistry, Sulfonic Acid Derivatives, Materials Chemistry, Monomers
Other Product Names / Synonyms: [3-(2-Nitrobenzenesulfonamido)propyl]carbamic Acid Allyl Ester

Formula: C₁₃H₁₇N₃O₆S
MW: 343.35 g/mol
CAS Number: 312283-45-7

Order Now

Specification Test Results

Purity/Analysis Method 98.0% (HPLC)

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR