(1S,2S)-2-(Naphthalene-2,3-dicarboximido)cyclohexanecarboxylic acid ≥98.0% (by HPLC, titration analysis)
Supplier: TCI America
Synonyms:
N-[(1S,2S)-2-Carboxycyclohexyl]naphthalene-2,3-dicarboximide
CAS Number: 642995-16-2
Molecular Formula: C19H17NO4
Molecular Weight: 323.35
Purity/Analysis Method:
Form: Crystal
Specific rotation [a]20/D: 34 deg (C=0.3, THF)
Product Categories: Analytical Science, e.e. Determination (HPLC Labeling Reagents), e.e. / Absolute Configuration Determination (NMR), Fluorescence Detection (HPLC Labeling Reagents), Fluorescence Detection (HPLC Labeling Reagents), UV Detection (HPLC Labeling Reagents), UV
Other Product Names / Synonyms: N-[(1S,2S)-2-Carboxycyclohexyl]naphthalene-2,3-dicarboximide
Formula:
C₁₉H₁₇NO₄ MW: 323.35 g/mol |
CAS Number:
642995-16-2 |
Specification Test Results
Purity/Analysis Method |
|
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...