Potassium perfluorobutane-1-sulfonate ≥98.0% (by titrimetric analysis)

Supplier: TCI America

Synonyms: Perfluoro-1-butanesulfonic acid potassium saltPerfluoro-1-butanesulphonic acid potassium saltPotassium Nonafluoro-1-butanesulphonatePotassium nonafluoro-1-butanesulphonatePotassium nonafluoro-1-butanesulfonatePotassium Perfluoro-1-butanesulphonatePotassium Nonafluoro-1-butanesulfonateNonafluoro-1-butanesulphonic acid potassium saltPotassium Perfluoro-1-butanesulfonateNonafluoro-1-butanesulfonic acid potassium salt

N0711-25G
TCN0711-25GEA 129.38 USD
TCN0711-25G
Potassium perfluorobutane-1-sulfonate ≥98.0% (by titrimetric analysis)
Potassium perfluorobutane-1-sulphonate

CAS Number: 29420-49-3
MDL Number: MFCD00007515
Molecular Formula: C4HF9O3S
Molecular Weight: 338.18
Purity/Analysis Method: 98.0% (T)
Form: Crystal


Product Categories: Synthetic Organic Chemistry, Fluorous Compounds, Materials Chemistry, Fluorosurfactants
Other Product Names / Synonyms: Nonafluoro-1-butanesulfonic Acid Potassium Salt, Perfluoro-1-butanesulfonic Acid Potassium Salt, Potassium Perfluoro-1-butanesulfonate

https://us.vwr.com/stibo/web/std.lang.all/99/43/28989943.jpg

Order Now

Specification Test Results

Purity/Analysis Method 98.0% (T)

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR