Lithium perfluorobutane-1-sulfonate ≥95.0% (by titrimetric analysis)

Supplier: TCI America

Synonyms: Lithium nonafluorobutanesulfonateLithium Nonafluoro-1-butanesulfonateLithium Nonafluoro-1-butanesulphonateLithium nonafluorobutanesulphonate

N0710-25G
TCN0710-25GEA 135.97 USD
TCN0710-25G
Lithium perfluorobutane-1-sulfonate ≥95.0% (by titrimetric analysis)
Lithium perfluorobutane-1-sulphonate

CAS Number: 131651-65-5
MDL Number: MFCD03094160
Molecular Formula: C4HF9O3S
Molecular Weight: 306.03
Purity/Analysis Method: 95.0% (T)
Form: Crystal


Product Categories: Synthetic Organic Chemistry, Fluorous Compounds, Materials Chemistry, Secondary Battery Research Reagents, Fluorosurfactants
Other Product Names / Synonyms: Lithium Perfluoro-1-butanesulfonate, Nonafluoro-1-butanesulfonic Acid Lithium Salt, Perfluoro-1-butanesulfonic Acid Lithium Salt

https://us.vwr.com/stibo/web/std.lang.all/99/42/28989942.jpg

Order Now

Specification Test Results

Purity/Analysis Method 95.0% (T)

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR