1,2,9-Nonanetriol ≥97.0%

Supplier: TCI America

N0652-5G
TCN0652-005GEA 458 USD
TCN0652-005G
1,2,9-Nonanetriol ≥97.0%
1,2,9-Nonanetriol

CAS Number: 382631-42-7
MDL Number: MFCD01861292
Molecular Formula: C9H20O3
Molecular Weight: 176.26
Purity/Analysis Method: 97.0% (GC)
Form: Crystal
Melting point (°C): 55

Formula: C₉H₂₀O₃
Melting Pt: 55 °C
MDL Number: MFCD01861292
CAS Number: 382631-42-7
https://us.vwr.com/stibo/web/std.lang.all/98/57/28989857.jpg

Order Now

Specification Test Results

Purity/Analysis Method 97.0% (GC)

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR