TCIAN0623

N0623-25G
TCN0623-025GEA 138.44 USD
TCN0623-025G
TCIAN0623
CAS Number: 4944-60-9
MDL Number: MFCD00450895
Molecular Formula: C13H24O4
Molecular Weight: 244.33
Purity/Analysis Method: 98.0% (GC)
Form: Clear Liquid
Boiling point (°C): 161
Specific Gravity (20/20): 0.98

Other Product Names / Synonyms: Nonamethylene Diacetate, 1,9-Nonanediol Diacetate
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR