3-Nitrophthalhydrazide ≥97.0% (by titrimetric analysis)
Supplier: TCI America
Synonyms:
3-Nitrophthalic Hydrazide
, 5-Nitro-2,3-dihydrophthalazine-1,4-dione
, 2,3-Dihydro-5-nitro-1,4-phthalazinedione
, 3-Nitrophthalhydrazide
N0622-5G
N0622-25G
TCN0622-025GEA
245.63
USD
TCN0622-025G
TCN0622-5G
3-Nitrophthalhydrazide ≥97.0% (by titrimetric analysis)
3-Nitrophthalhydrazide
CAS Number: 3682-15-3
MDL Number: MFCD00006889
Molecular Formula: C8H5N3O4
Molecular Weight: 207.15
Purity/Analysis Method:
Form: Crystal
Other Product Names / Synonyms: 2,3-Dihydro-5-nitro-1,4-phthalazinedione, 3-Nitrophthalhydrazide
Formula:
C₈H₅N₃O₄ MW: 207.1430 g/mol |
MDL Number:
MFCD00006889 CAS Number: 3682-15-3 |
Specification Test Results
Purity/Analysis Method |
|
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...