1-(4-Nitrophenyl)-1H-pyrrole ≥98.0%

Supplier: TCI America

Synonyms: 1-(4-Nitrophenyl)pyrrole

N0584-1G
TCN0584-001GEA 118 USD
TCN0584-001G
1-(4-Nitrophenyl)-1H-pyrrole ≥98.0%
1-(4-Nitrophenyl)-1H-pyrrole

CAS Number: 4533-42-0
MDL Number: MFCD00119340
Molecular Formula: C10H8N2O2
Molecular Weight: 188.19
Purity/Analysis Method: 98.0% (GC)
Form: Crystal

Formula: C10H8N2O2
MW: 188.1800 g/mol
MDL Number: MFCD00119340
CAS Number: 4533-42-0
https://us.vwr.com/stibo/web/std.lang.all/97/67/28989767.jpg

Order Now

Specification Test Results

Purity/Analysis Method 98.0% (GC)

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR