N-(1-Naphthalenesulfonyl)-L-phenylalanyl chloride ≥98.0% (by titrimetric analysis)

N0581-5G
TCN0581-005GEA 244.2 USD
TCN0581-005G
N-(1-Naphthalenesulfonyl)-L-phenylalanyl chloride ≥98.0% (by titrimetric analysis)
N-(1-Naphthalenesulfonyl)-L-phenylalanyl chloride

CAS Number: 146864-62-2
MDL Number: MFCD00191468
Molecular Formula: C19H16ClNO3S
Molecular Weight: 373.85
Purity/Analysis Method: 98.0% (T)
Form: Crystal
Melting point (°C): 104
Specific rotation [a]20/D: 19 deg (C=2, Dioxane)

Formula: C₁₉H₁₆ClNO₃S
MW: 373.85 g/mol
Melting Pt: 104 °C
MDL Number: MFCD00191468
CAS Number: 146864-62-2
https://us.vwr.com/stibo/web/std.lang.all/97/65/28989765.jpg

Order Now

Specification Test Results

Purity/Analysis Method 98.0% (T)

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR