N-(4-Nitrophenylsulfonyl)-L-phenylalanine ≥98.0% (by titrimetric analysis)

Supplier: TCI America

N0580-5G
TCN0580-005GEA 165.37 USD
TCN0580-005G
N-(4-Nitrophenylsulfonyl)-L-phenylalanine ≥98.0% (by titrimetric analysis)
N-(4-Nitrophenylsulfonyl)-L-phenylalanine

CAS Number: 64501-87-7
MDL Number: MFCD00191470
Molecular Formula: C15H14N2O6S
Molecular Weight: 350.35
Purity/Analysis Method: 98.0% (T)
Form: Crystal
Melting point (°C): 169
Specific rotation [a]20/D: -19 deg (C=3, Acetone)

Formula: C₁₅H₁₄N₂O₆S
Melting Pt: 169 °C
MDL Number: MFCD00191470
CAS Number: 64501-87-7
https://us.vwr.com/stibo/web/std.lang.all/97/64/28989764.jpg

Order Now

Specification Test Results

Purity/Analysis Method 98.0% (T)

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR