N-(1-Naphthalenesulfonyl)-L-phenylalanine ≥98.0% (by titrimetric analysis)

Supplier: TCI America

N0579-5G
TCN0579-005GEA 217.12 USD
TCN0579-005G
N-(1-Naphthalenesulfonyl)-L-phenylalanine ≥98.0% (by titrimetric analysis)
N-(1-Naphthalenesulfonyl)-L-phenylalanine

CAS Number: 90291-46-6
MDL Number: MFCD00191467
Molecular Formula: C19H17NO4S
Molecular Weight: 355.41
Purity/Analysis Method: 98.0% (T)
Form: Crystal
Melting point (°C): 141
Specific rotation [a]20/D: -33 deg (C=3, Acetone)

Formula: C₁₉H₁₇NO₄S
Melting Pt: 141 °C
MDL Number: MFCD00191467
CAS Number: 90291-46-6
https://us.vwr.com/stibo/web/std.lang.all/97/63/28989763.jpg

Order Now

Specification Test Results

Purity/Analysis Method 98.0% (T)

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR