Acid green A ≥97.0% (by HPLC)

Supplier: TCI America

A1905-25G
TCA1905-25GEA 213 USD
TCA1905-25G
Acid green A ≥97.0% (by HPLC)
Acid green A

CAS Number: 93942-43-9
Molecular Formula: C29H36N2O6S2
Molecular Weight: 610.83
Purity/Analysis Method: 97.0% (HPLC)
Form: Crystal

Formula: C₂₉H₃₆N₂O₆S₂
CAS Number: 93942-43-9
https://us.vwr.com/stibo/web/std.lang.all/00/00/28930000.jpg

Order Now

Specification Test Results

Purity/Analysis Method 97.0% (HPLC)

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR