(S)-(-)-N-(5-Nitro-2-pyridyl)alaninol ≥98.0% (by titrimetric analysis)

Supplier: TCI America

N0547-100MG
TCN0547-100MGEA 30.43 USD
TCN0547-100MG
(S)-(-)-N-(5-Nitro-2-pyridyl)alaninol ≥98.0% (by titrimetric analysis)
(S)-(-)-N-(5-Nitro-2-pyridyl)alaninol

CAS Number: 115416-52-9
MDL Number: MFCD00145287
Molecular Formula: C8H11N3O3
Molecular Weight: 197.19
Purity/Analysis Method: 98.0% (T)
Form: Crystal
Melting point (°C): 100

Formula: C₈H₁₁N₃O₃
MDL Number: MFCD00145287
CAS Number: 115416-52-9

Order Now

Specification Test Results

Purity/Analysis Method 98.0% (T)

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR