Monomethyl-5-norbornene-2,3-dicarboxylate ≥99.0% (by titrimetric analysis)

Synonyms: Methyl Hydrogen 5-Norbornene-2,3-dicarboxylate3-(Methoxycarbonyl)bicyclo[2.2.1]hept-5-ene-2-carboxylic acid5-Norbornene-2,3-dicarboxylic Acid Monomethyl Ester

N0525-5G N0525-25G
TCN0525-5GEA 22.44 USD
TCN0525-5G TCN0525-025G
Monomethyl-5-norbornene-2,3-dicarboxylate ≥99.0% (by titrimetric analysis)
Monomethyl-5-norbornene-2,3-dicarboxylate

CAS Number: 36897-94-6
MDL Number: MFCD00167743
Molecular Formula: C10H12O4
Molecular Weight: 196.20
Purity/Analysis Method: 99.0% (T)
Form: Crystal
Melting point (°C): 100


Other Product Names / Synonyms: 5-Norbornene-2,3-dicarboxylic Acid Monomethyl Ester, Methyl Hydrogen 5-Norbornene-2,3-dicarboxylate

Formula: C₁₀H₁₂O₄
MW: 196.20 g/mol
Melting Pt: 100 °C
MDL Number: MFCD00167743
CAS Number: 36897-94-6

Order Now

Specification Test Results

Purity/Analysis Method 99.0% (T)

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR