10,12-Nonacosadiynoic acid ≥97.0% (by titrimetric analysis)
Supplier: TCI America
N0514-100MG
TCN0514-100MGEA
351
USD
TCN0514-100MG
10,12-Nonacosadiynoic acid ≥97.0% (by titrimetric analysis)
10,12-Nonacosadiynoic acid
CAS Number: 66990-34-9
MDL Number: MFCD00060119
Molecular Formula: C29H50O2
Molecular Weight: 430.72
Purity/Analysis Method:
Form: Crystal
Melting point (°C): 72
Storage Temperature: 0-10°C
Formula:
C₂₉H₅₀O₂ Melting Pt: 72 °C |
MDL Number:
MFCD00060119 CAS Number: 66990-34-9 |
Specification Test Results
Purity/Analysis Method |
|
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...