4-(1-Butylpentyl)pyridine ≥98.0% (by GC, titration analysis)

Supplier: TCI America

Synonyms: 4-(5-Nonyl)pyridine5-(4-Pyridyl)nonane

N0496-5ML
TCN0496-005MLEA 152.25 USD
TCN0496-005ML
4-(1-Butylpentyl)pyridine ≥98.0% (by GC, titration analysis)
4-(1-Butylpentyl)pyridine

CAS Number: 2961-47-9
MDL Number: MFCD00010710
Molecular Formula: C14H23N
Molecular Weight: 205.35
Purity/Analysis Method: 98.0% (GC,T)
Form: Clear Liquid
Boiling point (°C): 267
Flash Point (°C): 110
Specific Gravity (20/20): 0.89


Other Product Names / Synonyms: 5-(4-Pyridyl)nonane

Formula: C₁₄H₂₃N
MW: 205.34 g/mol
Boiling Pt: 267 °C
Density: 0.89 (20 °C)
Flash Pt: 110 °C
MDL Number: MFCD00010710
CAS Number: 2961-47-9
EINECS: 220-993-6

Order Now

Specification Test Results

Purity/Analysis Method 98.0% (GC,T)

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR