2-Nitrophenetole ≥98.0%

Supplier: TCI America

Synonyms: 1-Ethoxy-2-nitrobenzene

N0489-25G
TCN0489-025GEA 164.35 USD
TCN0489-025G
2-Nitrophenetole ≥98.0%
2-Nitrophenetole

CAS Number: 610-67-3
MDL Number: MFCD00053603
Molecular Formula: C8H9NO3
Molecular Weight: 167.16
Purity/Analysis Method: 98.0% (GC)
Form: Clear Liquid
Boiling point (°C): 275
Freezing point (°C): 4
Specific Gravity (20/20): 1.19


Other Product Names / Synonyms: 1-Ethoxy-2-nitrobenzene

Formula: C₈H₉NO₃
MW: 167.16 g/mol
Boiling Pt: 275 °C
Density: 1.19 (20 °C)
MDL Number: MFCD00053603
CAS Number: 610-67-3
https://us.vwr.com/stibo/web/std.lang.all/96/47/28989647.jpg

Order Now

Specification Test Results

Purity/Analysis Method 98.0% (GC)

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR