syn-4-Nitrobenzaldehyde oxime ≥85.0% [Deprotecting Agent]

Supplier: TCI America
Warning

Synonyms: syn-4-Nitrobenzaldoxime

N0487-25G
TCN0487-025GEA 167 USD
TCN0487-025G
syn-4-Nitrobenzaldehyde oxime ≥85.0% [Deprotecting Agent]
syn-4-Nitrobenzaldehyde oxime
Formula: C₇H₆N₂O₃
MW: 166.14 g/mol
Melting Pt: 126 °C
MDL Number: MFCD00151329
CAS Number: 3717-19-9
UN: 2811
ADR: 6.1,III
https://us.vwr.com/stibo/web/std.lang.all/96/46/28989646.jpg

Order Now

Specification Test Results

Purity/Analysis Method 85.0% (GC)

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR