4'-Amino-3'-(trifluoromethyl)acetanilide ≥98.0% (by titrimetric analysis)

Supplier: TCI America

A1895-5G
TCA1895-5GEA 30.43 USD
TCA1895-5G
4'-Amino-3'-(trifluoromethyl)acetanilide ≥98.0% (by titrimetric analysis)
4'-Amino-3'-(trifluoromethyl)acetanilide

CAS Number: 1579-89-1
Molecular Formula: C9H9F3N2O
Molecular Weight: 218.18
Purity/Analysis Method: 98.0% (T)
Form: Crystal
Melting point (°C): 119


Product Categories: Synthetic Organic Chemistry, Fluorinated Building Blocks
Other Product Names / Synonyms: 5-Acetamido-2-aminobenzotrifluoride, 4-Acetamido-2-(trifluoromethyl)aniline

https://us.vwr.com/stibo/web/std.lang.all/99/82/28929982.jpg

Order Now

Specification Test Results

Purity/Analysis Method 98.0% (T)

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR