2-Propoxynaphthalene

Supplier: TCI America

Synonyms: 2-Naphthyl Propyl Ether

N0432-25G
TCN0432-025GEA 271 USD
TCN0432-025G
2-Propoxynaphthalene
2-Propoxynaphthalene

CAS Number: 19718-45-7
MDL Number: MFCD00021610
Molecular Formula: C13H14O
Molecular Weight: 186.25
Form: Crystal
Color: White
Boiling point (°C): 305
Melting point (°C): 40
Storage Temperature: 0-10°C


Other Product Names / Synonyms: 2-Naphthyl Propyl Ether

Formula: C₁₃H₁₄O
MW: 186.25 g/mol
Boiling Pt: 305 °C
Melting Pt: 40 °C
MDL Number: MFCD00021610
CAS Number: 19718-45-7
https://us.vwr.com/stibo/web/std.lang.all/95/46/28989546.jpg

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR