cis-4-Nonene ≥95.0%

Supplier: TCI America

N0384-5ML
TCN0384-005MLEA 292.06 USD
TCN0384-005ML
cis-4-Nonene ≥95.0%
cis-4-Nonene

CAS Number: 10405-84-2
MDL Number: MFCD00067487
Molecular Formula: C9H18
Molecular Weight: 126.24
Purity/Analysis Method: 95.0% (GC)
Form: Clear Liquid
Boiling point (°C): 147
Flash Point (°C): 30
Specific Gravity (20/20): 0.74

Formula: C₉H₁₈
Boiling Pt: 147 °C
Density: 0,74 (20 °C)
Flash Pt: 30 °C
MDL Number: MFCD00067487
CAS Number: 10405-84-2

Order Now

Specification Test Results

Purity/Analysis Method 95.0% (GC)

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR