TCIAN0305

N0305-100MG
TCN0305-100MGEA 51 USD
TCN0305-100MG
TCIAN0305
[for Determination of Ca in Serum]
CAS Number: 6409-77-4
MDL Number: MFCD00078493
Molecular Formula: C14H9NO7S
Molecular Weight: 357.27
Form: Crystal
Color: Yellow

Other Product Names / Synonyms: Kernechtrot, NFR
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR