4-Nitrosodiphenylamine ≥97.0% (by HPLC)

Supplier: TCI America

N0263-25G
TCN0263-025GEA 244 USD
TCN0263-025G
4-Nitrosodiphenylamine ≥97.0% (by HPLC)
4-Nitrosodiphenylamine

CAS Number: 156-10-5
MDL Number: MFCD00002062
Molecular Formula: C12H10N2O
Molecular Weight: 198.23
Purity/Analysis Method: 97.0% (HPLC)
Form: Crystal
Melting point (°C): 145

Formula: C₁₂H₁₀N₂O
Melting Pt: 145 °C
MDL Number: MFCD00002062
CAS Number: 156-10-5
https://us.vwr.com/stibo/web/std.lang.all/93/45/28989345.jpg

Order Now

Specification Test Results

Purity/Analysis Method 97.0% (HPLC)

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR