Acid green 27 ≥75.0% (by HPLC)
Supplier: TCI America
A1863-25G
TCA1863-25GEA
112
USD
TCA1863-25G
Acid green 27 ≥75.0% (by HPLC)
Acid green 27
CAS Number: 6408-57-7
MDL Number: MFCD00001196
Molecular Formula: C34H34N2O8S2
Molecular Weight: 706.74
Purity/Analysis Method:
Form: Crystal
Melting point (°C): 260
Lambda max.: 644 nm (H2O)
Formula:
C₃₄H₃₄N₂O₈S₂ Melting Pt: 260 °C |
MDL Number:
MFCD00001196 CAS Number: 6408-57-7 |
Specification Test Results
Purity/Analysis Method |
|
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...