5-Nitroguaiacol sodium salt ≥98.0% (by titrimetric analysis)

Supplier: TCI America

Synonyms: 2-Methoxy-5-nitrophenol Sodium Salt

N0205-25G
TCN0205-025GEA 200.88 USD
TCN0205-025G
5-Nitroguaiacol sodium salt ≥98.0% (by titrimetric analysis)
5-Nitroguaiacol sodium salt

CAS Number: 67233-85-6
MDL Number: MFCD00070570
Molecular Formula: C7H7NO4
Molecular Weight: 191.12
Purity/Analysis Method: 98.0% (T)
Form: Crystal


Other Product Names / Synonyms: 2-Methoxy-5-nitrophenol Sodium Salt

Formula: C₇H₇NO₄
MW: 191.12 g/mol
MDL Number: MFCD00070570
CAS Number: 67233-85-6
https://us.vwr.com/stibo/web/std.lang.all/92/95/28989295.jpg

Order Now

Specification Test Results

Purity/Analysis Method 98.0% (T)

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR