4-Amino-3,5-xylenol ≥98.0% (by GC, titration analysis)

Supplier: TCI America

Synonyms: 4-Amino-3,5-dimethylphenol

A1860-25G
TCA1860-25GEA 138 USD
TCA1860-25G
4-Amino-3,5-xylenol ≥98.0% (by GC, titration analysis)
4-Amino-3,5-xylenol

CAS Number: 3096-70-6
Molecular Formula: C8H11NO
Molecular Weight: 137.18
Purity/Analysis Method: 98.0% (GC,T)
Form: Crystal
Melting point (°C): 181


Other Product Names / Synonyms: 4-Amino-3,5-dimethylphenol, 4-Hydroxy-2,6-dimethylaniline

Formula: C₈H₁₁NO
MW: 137.1800 g/mol
Melting Pt: 181 °C
MDL Number: MFCD01741459
CAS Number: 3096-70-6
https://us.vwr.com/stibo/web/std.lang.all/99/64/28929964.jpg

Order Now

Specification Test Results

Purity/Analysis Method 98.0% (GC,T)

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR