3-Nitroaniline hydrochloride ≥98.0% (by HPLC, titration analysis)

Supplier: TCI America

N0120-25G
TCN0120-025GEA 82 USD
TCN0120-025G
3-Nitroaniline hydrochloride ≥98.0% (by HPLC, titration analysis)
3-Nitroaniline hydrochloride

CAS Number: 33240-96-9
MDL Number: MFCD00050693
Molecular Formula: C6H6N2O2
Molecular Weight: 174.58
Purity/Analysis Method: 98.0% (HPLC,T)
Form: Crystal

Formula: C₆H₇ClN₂O₂
MW: 174.5900 g/mol
MDL Number: MFCD00050693
CAS Number: 33240-96-9
UN: 1661
ADR: 6.1,II
https://us.vwr.com/stibo/web/std.lang.all/92/20/28989220.jpg

Order Now

Specification Test Results

Purity/Analysis Method 98.0% (HPLC,T)

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR