Dipotassium-2-naphthol-6,8-disulfonate hydrate ≥98.0% (by HPLC, titration analysis)
Supplier: TCI America
Synonyms:
G acid Dipotassium salt
, 7-Hydroxy-1,3-naphthalenedisulfonic acid Dipotassium salt
, 7-Hydroxy-1,3-naphthalenedisulphonic acid Dipotassium salt
, 2-Naphthol-6,8-disulfonic acid Dipotassium salt
, 2-Naphthol-6,8-disulphonic acid Dipotassium salt
, Dipotassium 7-Hydroxy-1,3-naphthalenedisulphonate
, Dipotassium 7-Hydroxy-1,3-naphthalenedisulfonate
CAS Number: 842-18-2
MDL Number: MFCD00066361
Molecular Formula: C10H8O7S2
Molecular Weight: 380.47
Purity/Analysis Method:
Form: Crystal
Other Product Names / Synonyms: Dipotassium 7-Hydroxy-1,3-naphthalenedisulfonate, 2-Naphthol-6,8-disulfonic Acid Dipotassium Salt, G Acid Dipotassium Salt, 7-Hydroxy-1,3-naphthalenedisulfonic Acid Dipotassium Salt
Formula:
C₁₀H₈O₇S₂ MW: 380.47 g/mol |
MDL Number:
MFCD00066361 CAS Number: 842-18-2 |
Specification Test Results
Purity/Analysis Method |
|
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...