1-Naphthohydroxamic acid ≥98.0% (by titrimetric analysis)

Supplier: TCI America

Synonyms: N-Hydroxynaphthalene-1-carboxamideN-Hydroxy-1-naphthamide

N0023-1G
TCN0023-001GEA 417 USD
TCN0023-001G
1-Naphthohydroxamic acid ≥98.0% (by titrimetric analysis)
1-Naphthohydroxamic acid

CAS Number: 6953-61-3
MDL Number: MFCD00059546
Molecular Formula: C11H9NO2
Molecular Weight: 187.20
Purity/Analysis Method: 98.0% (T)
Form: Crystal


Other Product Names / Synonyms: N-Hydroxynaphthalene-1-carboxamide

Formula: C₁₁H₉NO₂
MW: 187.20 g/mol
MDL Number: MFCD00059546
CAS Number: 6953-61-3
https://us.vwr.com/stibo/web/std.lang.all/91/44/28989144.jpg

Order Now

Specification Test Results

Purity/Analysis Method 98.0% (T)

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR