4'-Aminooctanophenone ≥97.0% (by GC, titration analysis)

Supplier: TCI America

A1849-25G
TCA1849-25GEA 147 USD
TCA1849-25G
4'-Aminooctanophenone ≥97.0% (by GC, titration analysis)
4'-Aminooctanophenone

CAS Number: 63884-78-6
Molecular Formula: C14H21NO
Molecular Weight: 219.33
Purity/Analysis Method: 97.0% (GC,T)
Form: Crystal
Melting point (°C): 105


Other Product Names / Synonyms: 4-n-Octanoylaniline

Formula: C₁₄H₂₁NO
Melting Pt: 105 °C
CAS Number: 63884-78-6
https://us.vwr.com/stibo/web/std.lang.all/99/45/28929945.jpg

Order Now

Specification Test Results

Purity/Analysis Method 97.0% (GC,T)

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR