3-Methoxypyridazine ≥97.0% (by GC, titration analysis)

Supplier: TCI America

M2078-1G M2078-5G
TCM2078-1GEA 80.15 USD
TCM2078-1G TCM2078-5G
3-Methoxypyridazine ≥97.0% (by GC, titration analysis)
3-Methoxypyridazine

CAS Number: 19064-65-4
MDL Number: MFCD00234098
Molecular Formula: C5H6N2O
Molecular Weight: 110.12
Purity/Analysis Method: 97.0% (GC,T)
Form: Clear Liquid
Boiling point (°C): 87
Specific Gravity (20/20): 1.15
Lambda max.: 265 nm (H2O)

Formula: C₅H₆N₂O
MW: 110.1139 g/mol
Boiling Pt: 87 °C
Density: 1.15 (20 °C)
MDL Number: MFCD00234098
CAS Number: 19064-65-4
https://us.vwr.com/stibo/web/std.lang.all/80/34/28988034.jpg

Order Now

Specification Test Results

Purity/Analysis Method 97.0% (GC,T)

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR