9-Mesityl-2,7,10-trimethylacridinium perchlorate ≥98.0% (by HPLC)
Supplier: TCI America
M2072-1G
TCM2072-1GEA
497.14
USD
TCM2072-1G
9-Mesityl-2,7,10-trimethylacridinium perchlorate ≥98.0% (by HPLC)
9-Mesityl-2,7,10-trimethylacridinium perchlorate
CAS Number: 1216909-33-9
Molecular Formula: C25H26ClNO4
Molecular Weight: 439.94
Purity/Analysis Method:
Form: Crystal
Storage Temperature: 0-10°C
Formula:
C₂₅H₂₆ClNO₄ |
CAS Number:
1216909-33-9 |
Specification Test Results
Purity/Analysis Method |
|
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...