Allyl-4-hydroxybenzoate ≥98.0% (by GC, titration analysis)

Supplier: TCI America

Synonyms: 4-Hydroxybenzoic Acid Allyl EsterAllylparaben

A1846-25G
TCA1846-25GEA 166.41 USD
TCA1846-25G
Allyl-4-hydroxybenzoate ≥98.0% (by GC, titration analysis)
Allyl-4-hydroxybenzoate

CAS Number: 18982-18-8
Molecular Formula: C10H10O3
Molecular Weight: 178.19
Purity/Analysis Method: 98.0% (GC,T)
Form: Crystal
Melting point (°C): 103


Product Categories: Materials Chemistry, Monomers
Other Product Names / Synonyms: Allylparaben, 4-Hydroxybenzoic Acid Allyl Ester

Formula: C₁₀H₁₀O₃
MW: 178.19 g/mol
Melting Pt: 103 °C
CAS Number: 18982-18-8
https://us.vwr.com/stibo/web/std.lang.all/99/42/28929942.jpg

Order Now

Specification Test Results

Purity/Analysis Method 98.0% (GC,T)

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR