2-Methyl-1-(p-tolyl)-2-propanol ≥98.0%

Synonyms: alpha,alpha,4-Trimethylphenethyl Alcohol1,1-Dimethyl-2-(p-tolyl)ethanol

M2019-5G M2019-25G
TCM2019-5GEA 51 USD
TCM2019-5G TCM2019-25G
2-Methyl-1-(p-tolyl)-2-propanol ≥98.0%
2-Methyl-1-(p-tolyl)-2-propanol

CAS Number: 20834-59-7
MDL Number: MFCD00029997
Molecular Formula: C11H16O
Molecular Weight: 164.25
Purity/Analysis Method: 98.0% (GC)
Form: Crystal
Boiling point (°C): 112
Melting point (°C): 42


Other Product Names / Synonyms: 1,1-Dimethyl-2-(p-tolyl)ethanol, alpha,alpha,4-Trimethylphenethyl Alcohol

Formula: C₁₁H₁₆O
MW: 164.25 g/mol
Boiling Pt: 112 °C
Melting Pt: 42 °C
MDL Number: MFCD00029997
CAS Number: 20834-59-7
https://us.vwr.com/stibo/web/std.lang.all/79/60/28987960.jpg

Order Now

Specification Test Results

Purity/Analysis Method 98.0% (GC)

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR