3-Methoxyphthalide ≥98.0%

Supplier: TCI America

Synonyms: 3-Methoxyisobenzofuran-1(3H)-one3-Methoxy-1(3H)-isobenzofuranone

M2007-5G
TCM2007-5GEA 173 USD
TCM2007-5G
3-Methoxyphthalide ≥98.0%
3-Methoxyphthalide

CAS Number: 4122-57-0
Molecular Formula: C9H8O3
Molecular Weight: 164.16
Purity/Analysis Method: 98.0% (GC)
Form: Crystal
Boiling point (°C): 146
Melting point (°C): 45
Storage Temperature: 0-10°C


Other Product Names / Synonyms: 3-Methoxy-1(3H)-isobenzofuranone

Formula: C₉H₈O₃
MW: 164.1580 g/mol
Boiling Pt: 146 °C
Melting Pt: 45 °C
CAS Number: 4122-57-0
https://us.vwr.com/stibo/web/std.lang.all/79/43/28987943.jpg

Order Now

Specification Test Results

Purity/Analysis Method 98.0% (GC)

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR