(S)-2-[(1-Methyl-2-pyrrolidinyl)methyl]isoindoline dihydrochloride ≥98.0% (by titrimetric analysis)
Supplier: TCI America
Synonyms:
(S)-2-(2-Isoindolinylmethyl)-1-methylpyrrolidine Dihydrochloride
M1995-1G
TCM1995-1GEA
180
USD
TCM1995-1G
(S)-2-[(1-Methyl-2-pyrrolidinyl)methyl]isoindoline dihydrochloride ≥98.0% (by titrimetric analysis)
(S)-2-[(1-Methyl-2-pyrrolidinyl)methyl]isoindoline dihydrochloride
Molecular Weight: 289.24
Purity/Analysis Method:
Form: Crystal
Specific rotation [a]20/D: -44 deg (C=1, H2O)
Molecular Formula: C14H20N2
Other Product Names / Synonyms: (S)-2-(2-Isoindolinylmethyl)-1-methylpyrrolidine Dihydrochloride
Specification Test Results
Purity/Analysis Method |
|
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...