Mono-2-O-(p-toluenesulfonyl)-γ-cyclodextrin ≥95.0% (by HPLC)

Supplier: TCI America

M1957-200MG M1957-1G
TCM1957-200MGEA 82.62 USD
TCM1957-200MG TCM1957-1G
Mono-2-O-(p-toluenesulfonyl)-γ-cyclodextrin ≥95.0% (by HPLC)
Mono-2-O-(p-toluenesulfonyl)-gamma-cyclodextrin

CAS Number: 97227-32-2
Molecular Formula: C55H86O42S
Molecular Weight: 1451.31
Purity/Analysis Method: 95.0% (HPLC)
Form: Crystal
Specific rotation [a]20/D: 148 deg (C=4, DMSO)

Formula: C₅₅H₈₆O₄₂S
CAS Number: 97227-32-2
https://us.vwr.com/stibo/web/std.lang.all/78/97/28987897.jpg

Order Now

Specification Test Results

Purity/Analysis Method 95.0% (HPLC)

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR