10-Methoxy-5H-dibenzo[b,f]azepine ≥98.0% (by HPLC, total nitrogen)
Supplier: TCI America
Synonyms:
10-Methoxy-2,2'-iminostilbene
M1895-25G
TCM1895-25GEA
115.66
USD
TCM1895-25G
10-Methoxy-5H-dibenzo[b,f]azepine ≥98.0% (by HPLC, total nitrogen)
10-Methoxy-5H-dibenzo[b,f]azepine
CAS Number: 4698-11-7
MDL Number: MFCD00101079
Molecular Formula: C15H13NO
Molecular Weight: 223.28
Purity/Analysis Method:
Form: Crystal
Melting point (°C): 127
Storage Temperature: 0-10°C
Other Product Names / Synonyms: 10-Methoxy-2,2'-iminostilbene
Formula:
C₁₅H₁₃NO MW: 223.28 g/mol Melting Pt: 127 °C |
MDL Number:
MFCD00101079 CAS Number: 4698-11-7 |
Specification Test Results
Purity/Analysis Method |
|
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...