8-Methoxy-4-methylbenzo[g]coumarin ≥98.0% (by HPLC)

Supplier: TCI America

Synonyms: 8-Methoxy-4-methyl-2H-naphtho[2,3-b]pyran-2-one

M1862-100MG
TCM1862-100MGEA 376 USD
TCM1862-100MG
8-Methoxy-4-methylbenzo[g]coumarin ≥98.0% (by HPLC)
8-Methoxy-4-methylbenzo[g]coumarin

CAS Number: 69169-71-7
Molecular Formula: C15H12O3
Molecular Weight: 240.26
Purity/Analysis Method: 98.0% (HPLC)
Form: Crystal
Melting point (°C): 221


Other Product Names / Synonyms: 8-Methoxy-4-methyl-2H-naphtho[2,3-b]pyran-2-one

Formula: C₁₅H₁₂O₃
MW: 240.26 g/mol
Melting Pt: 221 °C
CAS Number: 69169-71-7
https://us.vwr.com/stibo/web/std.lang.all/62/87/28976287.jpg

Order Now

Specification Test Results

Purity/Analysis Method 98.0% (HPLC)

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR