1-Methoxyphenazine ≥97.0% (by GC, titration analysis)
Supplier: TCI America
M1828-1G
TCM1828-1GEA
166.41
USD
TCM1828-1G
1-Methoxyphenazine ≥97.0% (by GC, titration analysis)
1-Methoxyphenazine
CAS Number: 2876-17-7
MDL Number: MFCD01794656
Molecular Formula: C13H10N2O
Molecular Weight: 210.24
Purity/Analysis Method:
Form: Crystal
Melting point (°C): 171
Formula:
C₁₃H₁₀N₂O Melting Pt: 171 °C |
MDL Number:
MFCD01794656 CAS Number: 2876-17-7 |
Specification Test Results
Purity/Analysis Method |
|
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...