(2-Methyl-3-nitrophenyl)acetic acid ≥96.0% (by GC, titration analysis)
Supplier: TCI America
Synonyms:
2-Methyl-3-nitrophenylacetic Acid
M1817-5G
TCM1817-5GEA
28.42
USD
TCM1817-5G
(2-Methyl-3-nitrophenyl)acetic acid ≥96.0% (by GC, titration analysis)
(2-Methyl-3-nitrophenyl)acetic acid
CAS Number: 23876-15-5
MDL Number: MFCD07782132
Molecular Formula: C9H9NO4
Molecular Weight: 195.17
Purity/Analysis Method:
Form: Crystal
Melting point (°C): 133
Other Product Names / Synonyms: (3-Nitro-o-tolyl)acetic Acid
Formula:
C₉H₉NO₄ MW: 195.17 g/mol Melting Pt: 133 °C |
MDL Number:
MFCD07782132 CAS Number: 23876-15-5 |
Specification Test Results
Purity/Analysis Method |
|
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...